Learn More
Thermo Scientific™ Maxima Hot Start Taq DNA Polymerase
Enhance the specificity, sensitivity, and yield of DNA amplification with this hot start DNA polymerase.
Manufacturer:  Thermo Scientific™ EP0603
 View more versions of this product
Catalog No. FEREP0603
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Maxima Hot Start Taq DNA Polymerase is a chemically modified recombinant Taq DNA polymerase. The enzyme is inactive at room temperature, avoiding extension of non-specifically annealed primers or primer dimers and providing higher specificity of DNA amplification. The functional activity of the enzyme is restored during a short 4 minute incubation at 95°C. The activated enzyme maintains the same functionality as Taq DNA polymerase. Maxima Hot Start Taq DNA Polymerase is also available in a master mix format.
Highlights
  • Four minute activation time
  • High PCR specificity and sensitivity
  • Room temperature PCR set up
Specifications
Specifications
1000U
Maxima™ Hot Start™ Taq DNA Polymerase
Hot start PCR; High yield amplification of targets up to 3kb; RT-PCR.
10 x 1.25mL of 10X Hot Start PCR Buffer, 10 x 1.25mL of 25mM MgCl2