Thermo Scientific™ MaxiMix™ II Vortex Mixer

Simultaneously mix the contents of up to four test tubes or small closed-top flasks with the Thermo Scientific™ MaxiMix™ II Vortex Mixer.

Manufacturer: Thermo Scientific™ M37615Q

 View more versions of this product

Catalog No. 12-814-5Q

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Height (Metric) Exterior 15.2cm
    Length (Metric) Exterior 17.7cm
    Width (Metric) Exterior 11.4cm
    Shipping Weight (English) 8 lb.
    Shipping Weight (Metric) 3.6kg
    Voltage 120V
    Platform Material Foam
    Hertz 50/60Hz
    Speed 100 to 3000rpm
    Amperage 0.75A
    Certifications/Compliance CSA approved
    Dimensions (L x W x H) Exterior 7 x 4.5 x 6 in. (17.7 x 11.4 x 15.2cm)
    Electrical Requirements 120V 50/60Hz
    Height (English) Exterior 6 in.
    Length (English) Exterior 7 in.
    Width (English) Exterior 4.5 in.

    Simultaneously mix the contents of up to four test tubes or small flasks with closed tops with the Thermo Scientific™ MaxiMix™ II Vortex Mixer. Featuring continuous operation and touch-on modes for gentle to vigorous mixing, it is excellent for mixing media in test tubes, mixing cytogenetic suspensions in centrifuge tubes, vortexing cell suspensions and vortexing drug extractions. It is ideal for enzymatic and RIA assay applications, atomic absorption sample preparation and Nelson's assay for reducing sugars.

    Features:

    • Continuous-run or touch-activated run modes
    • White, durable plastic housing resists acids and alkalines
    • Heavy-duty cast metal base with rubber feet assures stability and eliminates creep during use
    • Two mixing devices: Rubber single-cup tube holder and foam pad for mixing flasks or multiple tubes simultaneously

    Includes:

    • Single-tube cup
    • Foam pad
    • Power cord

    Recommended for:

    • Enzymatic and RIA assays
    • Vortexing cell suspensions
    • Drug extractions
    • Atomic absorption sample preparation
    • Nelson's assay for reducing sugars

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit