Learn More
Eppendorf™ Maxitips S Valve
Valve for Maxitips ensures drip-free dispensing of liquids with a high vapor pressure.
Supplier:  Eppendorf™ 022291459
Catalog No. 05-414-111
Add to cart
This item is not returnable. View return policy
Description
Description
  • Use Maxitips S in combination with the Eppendorf Maxitip to aspirate between 2.5 mL and 10 mL liquid from high, narrow-mouth vessels with the air cushion principle
  • Valve for Maxitips ensures drip-free dispensing of liquids with a high vapour pressure
  • Graduation on Maxitips ensures accurate dispensing of non-aqueous liquids
Specifications
Specifications
2.5 to 10 mL
100/Pk.
Eppendorf™ Maxitip™ S
Pipetting Aqueous Solutions, Extraction of Liquids from Deep Receptacles, Transfer of Organic Liquids, Solvents with High Vapor Pressure, Liquids with High Density, and Viscous Liquids
Valve