Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MaxQ™ 2508 Dual-Action Orbital Reciprocating Shaker

Facilitate repeatable results with this shaker, ideal for growing bacteria, dissolution studies and staining of electrophoresis gels and chemical extractions.

$3270.00

Specifications

Certifications/Compliance UL, cUL
Control Analog
Dimensions (L x W x H) Exterior 17.4 x 13.5 x 7 in. (44.2 x 34.3 x 17.8cm)
Humidity Range 20 to 80%
Load Bearing Capacity 15 lb. (6.8kg)
View More Specs

Products 1
Catalog Number Mfr. No. Electrical Requirements Certifications/Compliance Price Quantity & Availability  
Catalog Number Mfr. No. Electrical Requirements Certifications/Compliance Price Quantity & Availability  
11-675-150
Accessories View Documents
Thermo Scientific™
SHKA2508
120V 50/60Hz UL, cUL
Each for $3,270.00
Add to cart
 
Description

Description

Facilitate repeatable results with Thermo Scientific™ MaxQ™ 2508 Dual-Action Orbital Reciprocating Shakers, ideal for growing bacteria, dissolution studies and staining of electrophoresis gels and chemical extractions. An added reciprocating function, analog operating system and choice of three platforms make this an affordable shaker with increased capacity for a wide range of applications. Maximize your orbital reciprocating shaking capacity by choosing your platform with pre-installed clamps.

Features:
  • Variable speed control allows gentle to vigorous agitation
  • Built-in timer facilitates repeatable results
  • Control lever changes the motion while the shaker is stationary or in operation.
  • Shaking motion: back-and-forth, with fixed stroke angle of 0.4 in. (1.2cm) for shaking of separatory funnels, or fixed circular motion of 0.4 in. (1.2cm) for growing bacteria and yeast
  • Can be used in incubators, warm rooms, environmental chambers and refrigerators
  • Operates in temperature ranges of 4° to 40°C and humidity conditions of 20 to 80% noncondensing
  • Timed (1 to 60 min.) or continuous
  • Metal locking tab on speed control knob prevents increased or decreased speed if the knob is bumped

Requires:

  • Universal Platform (without clamps), or
  • Dedicated Platform (with pre-installed clamps) Clamps
Specifications

Specifications

UL, cUL
17.4 x 13.5 x 7 in. (44.2 x 34.3 x 17.8cm)
15 lb. (6.8kg)
DC
38 lb.
40 to 400rpm
1 to 60 min. or continuous
Analog
20 to 80%
MaxQ 2508
0.4 in. (1.2cm) circular or fixed stroke angle
17.2kg
4° to 40°C (Operating)
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit