Learn More
Thermo Scientific™ MaxQ™ 3000/5000 Dedicated Platforms with Clamps
Use these dedicated platforms with Thermo Scientific™ MaxQ™ 3000 and 5000 Shakers.
Manufacturer:  Thermo Scientific™ 359958
 View more versions of this product
Catalog No. 11-675-447
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Achieve maximum capacity if only using a single vessel size such as a flask with Thermo Scientific™ MaxQ™ 3000/5000 Dedicated Platforms with Clamps. Use these dedicated platforms with MaxQ™ 3000 and 5000 Shakers.
  • Clamps come installed to provide maximum capacity
Specifications
Specifications
MaxQ 3000 Shaker Only
24 in.
61cm
Erlenmeyer Flask
36 in.
91cm
15
MaxQ™ 3000 Shaker Only