Learn More
Thermo Scientific™ MaxQ™ HP Orbital Shakers Dedicated Platforms with Clamps
Maximize capacity when working with a single-size vessel by using HP Orbital Shakers Dedicated Platforms with pre-installed clamps.
$1191.00 - $3816.00
Specifications
Type Shaker Platform with Clamps
View More Specs
Products 32
Catalog Number Mfr. No. Clamps Compatibility For Use With Price Quantity & Availability  
Catalog Number Mfr. No. Clamps Compatibility For Use With Price Quantity & Availability  
238010
View Documents
Thermo Scientific™
238010
49 25mL Erlenmeyer Flask MaxQ HP 416/420
Each for $1,304.00
Add to cart
238011
View Documents
Thermo Scientific™
238011
49 50mL Erlenmeyer Flask MaxQ HP 416/420
Each for $1,531.00
Add to cart
238017
View Documents
Thermo Scientific™
238017
91 25mL Erlenmeyer Flask MaxQ HP 430/435/436/480/481
Each for $2,209.00
Add to cart
238018
View Documents
Thermo Scientific™
238018
91 50mL Erlenmeyer Flask MaxQ HP 430/435/436/480/481
Each for $2,188.00
Add to cart
238019
View Documents
Thermo Scientific™
238019
61 125mL Erlenmeyer Flask MaxQ HP 430/435/436/480/481
Each for $2,130.00
Add to cart
238022
View Documents
Thermo Scientific™
238022
15 1L Erlenmeyer Flask MaxQ HP 430/435/436/480/481
Each for $1,757.00
Add to cart
238024
View Documents
Thermo Scientific™
238024
6 4L Erlenmeyer Flask MaxQ HP 430/435/436/480/481
Each for $1,630.00
Add to cart
238025
View Documents
Thermo Scientific™
238025
4 6L Erlenmeyer Flask MaxQ HP 430/435/436/480/481
Each for $1,642.00
Add to cart
238029
View Documents
Thermo Scientific™
238029
30 500mL Erlenmeyer Flask MaxQ HP 430
Each for $2,386.00
Add to cart
238030
View Documents
Thermo Scientific™
238030
24 1L Erlenmeyer Flask MaxQ HP 430
Each for $2,291.00
Add to cart
238031
View Documents
Thermo Scientific™
238031
15 2L Erlenmeyer Flask MaxQ HP 430
Each for $2,282.00
Add to cart
238032
View Documents
Thermo Scientific™
238032
11 4L Erlenmeyer Flask MaxQ HP 430
Each for $2,603.00
Add to cart
238034
View Documents
Thermo Scientific™
238034
11 2800mL Fernbach Flask MaxQ HP 430
Each for $2,686.00
Add to cart
238036
View Documents
Thermo Scientific™
238036
80 25mL Erlenmeyer Flask MaxQ HP 416; Max. 400rpm
Each for $2,261.00
Add to cart
238037
View Documents
Thermo Scientific™
238037
80 50mL Erlenmeyer Flask MaxQ HP 416; Max. 400rpm
Each for $1,985.00
Add to cart
238038
View Documents
Thermo Scientific™
238038
40 125mL Erlenmeyer Flask MaxQ HP 416; Max. 400rpm
Each for $1,718.00
Add to cart
238039
View Documents
Thermo Scientific™
238039
35 250/300mL Erlenmeyer Flask MaxQ HP 416; Max. 400rpm
Each for $1,805.00
Add to cart
238040
View Documents
Thermo Scientific™
238040
20 500mL Erlenmeyer Flask MaxQ HP 416; Max. 400rpm
Each for $1,871.00
Add to cart
238043
View Documents
Thermo Scientific™
238043
5 4L Erlenmeyer Flask MaxQ HP 416; Max. 400rpm
Each for $1,480.00
Add to cart
238044
View Documents
Thermo Scientific™
238044
4 6L Erlenmeyer Flask MaxQ HP 416; Max. 400rpm
Each for $1,480.00
Add to cart
238045
View Documents
Thermo Scientific™
238045
5 2800mL Fernbach Flask MaxQ HP 416; Max. 400rpm
Each for $1,480.00
Add to cart
238046
View Documents
Thermo Scientific™
238046
4 4L Erlenmeyer Flask MaxQ HP 416
Each for $1,480.00
Add to cart
238047
View Documents
Thermo Scientific™
238047
2 6L Erlenmeyer Flask MaxQ HP 416
Each for $1,480.00
Add to cart
238048
View Documents
Thermo Scientific™
238048
4 2800mL Fernbach Flask MaxQ HP 416/420
Each for $1,256.00
Add to cart
238049
View Documents
Thermo Scientific™
238049
36 125mL Erlenmeyer Flask MaxQ HP 416/420
Each for $1,325.00
Add to cart
238051
View Documents
Thermo Scientific™
238051
30 250/300mL Erlenmeyer Flask MaxQ HP 430/435/436/480/481
Each for $2,118.00
Add to cart
238053
View Documents
Thermo Scientific™
238053
4 5L Erlenmeyer Flask MaxQ HP 416
Each for $1,191.00
Add to cart
238054
missing translation for 'viewDocuments'
Thermo Scientific™
238054
6 5L Erlenmeyer Flask MaxQ HP 430/435/436/480/481
Each for $1,920.00
Add to cart
238056
missing translation for 'viewDocuments'
Thermo Scientific™
238056
8 5L Erlenmeyer Flask MaxQ HP 430
Each for $2,271.00
Add to cart
238057
missing translation for 'viewDocuments'
Thermo Scientific™
238057
150 25mL Erlenmeyer Flask MaxQ HP 430
Each for $3,516.00
Add to cart
238058
missing translation for 'viewDocuments'
Thermo Scientific™
238058
150 50mL Erlenmeyer Flask MaxQ HP 430
Each for $3,816.00
Add to cart
238023
missing translation for 'viewDocuments'
Thermo Scientific™
238023
12 2L Erlenmeyer Flask MaxQ HP 430/435/436/480/481 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Maximize capacity when working with a single-size vessel by using Thermo Scientific™ HP Orbital Shakers Dedicated Platforms with pre-installed clamps.
Specifications
Specifications
Shaker Platform with Clamps