Learn More
Sklar InstrumentsSupplier Diversity Partner Mayo Dissecting Scissors, Sklar, 9 in., Blunt/Blunt, Straight, Smooth, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 901590
Mayo Dissecting Scissors, Sklar, 9 in., Blunt/Blunt, Straight, Smooth, Stainless Steel. Sklar Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved, in a variety of lengths, and have a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. Sklar also offers Mayo Dissecting Scissors with a selection of special features to meet procedure specifications and surgeon preferences. This product is straight with smooth, blunt/blunt tips and a length of 9 inches.
Catalog No. 50-143-6540