Learn More
Sklar InstrumentsSupplier Diversity Partner Mayo Gallstone Scoop, Sklar, 10-3/4 in., Double End, Silver Plated. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 321040
Mayo Gallstone Scoop, Sklar, 10-3/4 in., Double End, Silver Plated. Sklar's Mayo Gallstone Scoop is a rigid, double-ended gall bladder instrument with spoon shaped tips used for removing gallstones from the gall bladder. This product is slightly curved at each end and measures 10-3/4 inches overall.
Catalog No. 50-143-8180