Learn More
Sklar InstrumentsSupplier Diversity Partner Mayo-Hegar Needle Holder, Sklar, 11-1/2 in., Blunt, Straight, Cross-Serrated with Groove, Finger Rings, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 202911
Mayo-Hegar Needle Holder, Sklar, 11-1/2 in., Blunt, Straight, Cross-Serrated with Groove, Finger Rings, Stainless Steel. Sklar Mayo-Hegar Needle Holders are widely used in general surgery for holding medium to large, heavy suture needles. The heavy jaws have cross-serrations and a groove. This instrument is available in several lengths. The shorter length instrument may be used for procedures such as suturing the uterine cervix, while longer lengths may be used for deep tissue suturing. This product is straight and measures 11-1/2 inches.
Catalog No. 50-143-7493