Learn More
Sklar InstrumentsSupplier Diversity Partner TC Mayo-Hegar Needle Holder, Sklar, 6-1/4 in., Blunt, Straight, Serrated, Finger Rings, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 218111
TC Mayo-Hegar Needle Holder, Sklar, 6-1/4 in., Blunt, Straight, Serrated, Finger Rings, Tungsten Carbide / Stainless Steel. The Sklar TC Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures. Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procdures. Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws. Tungsten carbide inserts also maximize control and precision. This product is straight with serrated jaws, and an overall length of 6-1/4 inches.
Catalog No. 50-143-7790