Learn More
Sklar InstrumentsSupplier Diversity Partner Mayo Instrument Holder, Sklar, 5-1/2 in., Stainless Steel. The hanger shaped design holds one finger ring of an instrument handle. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 101772
Mayo Instrument Holder, Sklar, 5-1/2 in., Stainless Steel. The Mayo Instrument Holder provides an organized way to hang forceps or scissors for storage or sterilization. The hanger shaped design holds one finger ring of an instrument handle while allowing the other to hang open. This provides the greatest opening for sterilization ensuring the entire instrument is accessed during cleaning.
Catalog No. 50-143-7130