Learn More
Sklar InstrumentsSupplier Diversity Partner Mayo-Hegar Needle Holder, Sklar, 8 in., Blunt, Straight, Cross-Serrated with Groove, Finger Rings, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 202480
Mayo-Hegar Needle Holder, Sklar, 8 in., Blunt, Straight, Cross-Serrated with Groove, Finger Rings, Stainless Steel. The Sklar Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures. Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procdures, typically with larger, heavier suture needles because of it's wide jaws. Shorter lengths of the instrument may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. This product is straight with cross-serrated jaws featuring a central groove where the needle can be held, and a length of 8 inches.
Catalog No. 50-143-7476