Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Mayo-Hegar Needle Holder, Sklar, 7 in., Blunt, Straight, Serrated, Stainless Steel. PK/12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21853
Econo Mayo-Hegar Needle Holder, Sklar, 7 in., Blunt, Straight, Serrated, Stainless Steel. The Econo Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures. Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procdures. Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with cross-serrated jaws featuring a central groove with which the needle can be held, and a length of 7 inches. It is sold in boxes of 12.
Catalog No. 50-143-7830