Learn More
Mayo Scissors Products for Science Education
Can handle rough use.
Specifications
For Use With (Application) Surgical, Precision
Length (English) 5.5 in.
Length (Metric) 14cm
Tip Style Blunt/Blunt
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
S17339 Products for Science Education
View Documents
Medco Instruments
81300
N/A Straight This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
S17341 Products for Science Education
View Documents
Medco Instruments
81310
N/A Curved This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Long and strong
  • Made from stainless steel
  • Blunt point won't wear down
Specifications
Specifications
Surgical, Precision
5.5 in.
14cm
Blunt/Blunt