Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MB 0223

Dynamin-related GTPase DRP1 partial inhibitor

$209.00 - $881.00

Specifications

CAS 2247732-89-2
Chemical Name or Material 4-Methyl-1-((2-(piperidin-1-yl)thiazol-4-yl)methyl)-N-(4-(pyridin-3-yl)phenyl)-1H-pyrrole-2-carboxamide
Molecular Formula C26H27N5OS
Purity 98%
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
67-991-0
View Documents
Tocris Bioscience™
6799/10
10 mg
Each for $209.00
Add to cart
 
67-995-0
View Documents
Tocris Bioscience™
6799/50
50 mg
Each for $881.00
Add to cart
 
Description

Description

  • Dynamin-related GTPase DRP1 partial inhibitor (IC50 = 1.2 μM).
  • Selective for DRP1 over other dynamin-related GTPases, OPA1 and DYN1.
  • Increases mitochondrial DNA levels in mfn1- knockout MEFs deficient in mitochondrial fusion.
Specifications

Specifications

2247732-89-2
4-Methyl-1-((2-(piperidin-1-yl)thiazol-4-yl)methyl)-N-(4-(pyridin-3-yl)phenyl)-1H-pyrrole-2-carboxamide
C26H27N5OS
98%
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit