Learn More
Mopec MB800 Touch Screen Grossing Station Available on GSA/VA Contract for Federal Government customers only.
Designed to improve worker safety, increase operational versatility and provide more reliable equipment for your laboratory. Mopec™ MB800 Touch Screen Grossing Station focuses on ergonomics, workflow, and convenience.
Manufacturer:  Mopec MB800
Catalog No. 22-444-203
Add to cart
Description
Description
  • Industrial touch screen controller that monitors fan, air flow, optional formalin collection, optional formalin dispensing, optional filter monitoring
  • Hands free on/off control for lighting and fan system
  • Hot/cold water fixture with single lever control valves
  • Hand spray rinse
  • Back draft ventilation
  • Polyethylene cutting board
  • Flush inset dissecting perforated grid plates
  • 3 Recessed LED lights
  • 3 GFCI duplex electrical receptacles with USB 5V power outlets
  • Two Washable inch/centimeter ruler located at front edge of workstation
  • Variable position back splash mounting system with C-fold paper towel/glove dispenser, magnetic tool bar, and adjustable shelf
  • Camera ready integration
  • Built-in seismic anchor points
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Warranty and Services
1 Year Warranty
Specifications
Specifications
Elevating
152cm
195.58 - 226cm
22.86cm
Industrial touch screen
Back draft ventilation
91cm
35.56cm
45.72cm
3 GFCI duplex electrical receptacles with USB 5V power outlets