Learn More
Aat BioquestSupplier Diversity Partner mBCl [Monochlorobimane] *CAS 76421-73-3* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 635
Monochloromobimane (mBCl) is essentially nonfluorescent until reacted with a thiol group. It readily reacts with several low molecular weight thiols, including cysteine, glutathione, N-acetylcysteine, mercaptopurine, peptides and plasma thiols.
Catalog No. 50-195-2943