Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD1 Mouse anti-Human, Clone: CPTC-MBD1-1, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen MBD1
Clone CPTC-MBD1-1
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122024
View Documents
Novus Biologicals
NBP30727520UG
20 μg
Each for $138.37
Add to cart
 
NB122023
View Documents
Novus Biologicals
NBP30727510UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

MBD1 Monoclonal antibody specifically detects MBD1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

MBD1
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
CXXC-type zinc finger protein 3, methyl-CpG binding domain protein 1, methyl-CpG binding domain protein 1 isoform PCM1, methyl-CpG-binding domain protein 1, Methyl-CpG-binding protein MBD1, PCM1CXXC3RFT, Protein containing methyl-CpG-binding domain 1, the regulator of fibroblast growth factor 2 (FGF-2) transcription
Recombinant human MBD1 protein
Monoclonal
Protein A or G purified
CPTC-MBD1-1
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Chromatin Research
10 mM PBS with 0.05% BSA
4152
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit