Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5120803

This item is not returnable.
View return policy
Description
Positive test controls include: mouse heart, mouse kidney, rat heart. The target is usually found in the following locations: Nucleus.
MBD2 Polyclonal antibody specifically detects MBD2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
MBD2 | |
Polyclonal | |
Unconjugated | |
Mbd2 | |
demethylase; DKFZp586O0821; DMTase; MBD2; MBD2a; methyl-CpG binding domain protein 2; methyl-CpG-binding domain protein 2; Methyl-CpG-binding protein MBD2; NY-CO-41 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 202-411 of human MBD2 (NP_003918.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
3.1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Z2E1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17191, 680172 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title