Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen MBD2
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB126781
View Documents
Novus Biologicals
NBP310941100UL
100 μg
Each for $320.71
Add to cart
 
NB126782
View Documents
Novus Biologicals
NBP31094125UL
25 μg
Each for $106.19
Add to cart
 
Description

Description

MBD2 Polyclonal antibody specifically detects MBD2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

MBD2
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Purified
RUO
Human, Mouse
demethylase, DKFZp586O0821, DMTase, methyl-CpG binding domain protein 2, methyl-CpG-binding domain protein 2, Methyl-CpG-binding protein MBD2, NY-CO-41
The immunogen is a synthetic peptide directed towards the N terminal region of human MBD2 (NP_056647). Peptide sequence RAHPGGGRCCPEQEEGESAAGGSGAGGDSAIEQGGQGSALAPSPVSGVRR
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Unconjugated
Rabbit
Breast Cancer, Chromatin Research, Stem Cell Markers
PBS buffer, 2% sucrose
8932
Polyclonal
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit