Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBD3 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304590100UL
Description
MBD3 Polyclonal specifically detects MBD3 in Human, Mouse samples. It is validated for Western Blot.Specifications
MBD3 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Unconjugated | |
methyl-CpG binding domain protein 3, methyl-CpG-binding domain protein 3, Methyl-CpG-binding protein MBD3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-255 of human MBD3 (NP_001268383.1). | |
100 μL | |
Chromatin Research | |
53615 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'