Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBD4 Rabbit anti-Human, Mouse (Negative), Janelia Fluor 646, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP191189JF646
Description
MBD4 Polyclonal antibody specifically detects MBD4 in Human, Mouse (Negative) samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
MBD4 | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EC 3.2.2.-, G/T mismatch glycosylase, G/U mismatch glycosylase, MED1G/5-fluorouracil mismatch glycosylase with biphasic kinetics, methyl-CpG binding domain protein 4, methyl-CpG-binding domain protein 43,N(4)-ethenocytosine glycosylase, Methyl-CpG-binding endonuclease 1, Methyl-CpG-binding protein MBD4, Mismatch-specific DNA N-glycosylase, putative methyl-CpG binding protein | |
Rabbit | |
IgG | |
0.1 mL | |
Store at 4°C in the dark. | |
Polyclonal | |
Human, Mouse (Negative) |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Affinity Purified, Conjugated | |
MBD4 | |
A synthetic peptide made to an internal portion of the human MBD4 protein (between residues 200-250) [UniProt O95243] | |
Immunogen affinity purified | |
Base Excision Repair, Chromatin Research, DNA Repair, Epigenetics | |
Primary | |
8930.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title