Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBD4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen MBD4
Dilution Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation
Applications Western Blot, ChIP assay
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125590
View Documents
Novus Biologicals
NBP31034425UL
25 μg
Each for $106.19
Add to cart
 
NB125589
View Documents
Novus Biologicals
NBP310344100UL
100 μg
Each for $320.71
Add to cart
 
Description

Description

MBD4 Polyclonal antibody specifically detects MBD4 in Human samples. It is validated for Western Blot, ChIP assay
Specifications

Specifications

MBD4
Western Blot, ChIP assay
Purified
RUO
Human
EC 3.2.2.-, G/T mismatch glycosylase, G/U mismatch glycosylase, MED1G/5-fluorouracil mismatch glycosylase with biphasic kinetics, methyl-CpG binding domain protein 4, methyl-CpG-binding domain protein 43,N(4)-ethenocytosine glycosylase, Methyl-CpG-binding endonuclease 1, Methyl-CpG-binding protein MBD4, Mismatch-specific DNA N-glycosylase, putative methyl-CpG binding protein
The immunogen is a synthetic peptide directed towards the middle region of human MBD4 (NP_003916). Peptide sequence CSEQKTSGIINKFCSAKDSEHNEKYEDTFLESEEIGTKVEVVERKEHLHT
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation
Unconjugated
Rabbit
Base Excision Repair, Chromatin Research, DNA Repair, Epigenetics
PBS buffer, 2% sucrose
8930
Polyclonal
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit