Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBL Mouse anti-Chicken, Janelia Fluor 549, Clone: 6B11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP105085JF549

Catalog No. NBP15085549


Add to cart

Description

Description

MBL Monoclonal antibody specifically detects MBL in Chicken samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

MBL
6B11
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
MBL2
MBL purified from chicken plasma and adsorbed onto aluminum hydroxide
Protein A or G purified
Immunology
Primary
4153
Chicken
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
COLEC1collectin-1, Collectin-1, HSMBPC, Mannan-binding protein, Mannose-binding lectin, mannose-binding lectin (protein C) 2, soluble, mannose-binding lectin (protein C) 2, soluble (opsonic defect), mannose-binding protein C, MBLmannan-binding lectin, MBP, MBP1mannose-binding lectin 2, soluble (opsonic defect), MBP-C, MGC116832, MGC116833
Mouse
IgG1 Lambda
0.1mL
Store at 4°C in the dark.
Monoclonal
Mannan-binding lectin (MBL, chicken)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit