Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBL Mouse anti-Human, Alexa Fluor™ 532, Clone: 100, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223583AF532

Catalog No. NP22383A32


Add to cart

Description

Description

MBL Monoclonal antibody specifically detects MBL in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

MBL
100
Western Blot, ELISA
Conjugated, Purified
MBL2
MBL purified from human donor plasma and adsorbed onto aluminum hydroxide.
Protein A or G purified
Immunology
Primary
4153.0
Human
ELISA, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
COLEC1collectin-1, Collectin-1, HSMBPC, Mannan-binding protein, Mannose-binding lectin, mannose-binding lectin (protein C) 2, soluble, mannose-binding lectin (protein C) 2, soluble (opsonic defect), mannose-binding protein C, MBLmannan-binding lectin, MBP, MBP1mannose-binding lectin 2, soluble (opsonic defect), MBP-C, MGC116832, MGC116833
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Clone 1E2 is specific for MBL from human serum or plasma.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit