Learn More
Invitrogen™ MBNL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551603
Description
Immunogen sequence: AQMSSLGSFP MTPSIPANPP MAFNPYIPHP GMGLVPAELV PNTPVLIPGN PPLAMPGAVG PKLMRSDKLE VCREFQRGNC TRGENDCRYA HPTDASMIEA SDNTVTICMD YIKGRCSREK CKYFHPPAHL QARLKAAHHQ MNHSAAS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.
This gene encodes a member of the muscleblind-like family of proteins. The encoded protein may function in regulation of alternative splicing and may play a role in the pathophysiology of myotonic dystrophy. Alternatively spliced transcript variants have been described.
Specifications
MBNL3 | |
Polyclonal | |
Unconjugated | |
Mbnl3 | |
A530038J18Rik; AI661274; CHCR; Cys3His CCG1-required protein; E430034C16Rik; MBLX; MBLX39; Mbnl3; MBXL; muscleblind like splicing regulator 3; muscleblind-like 3; muscleblind-like 3 (Drosophila); muscleblind-like protein 3; muscleblind-like splicing regulator 3; muscleblind-like X-linked protein; Protein HCHCR; Protein MCHCR; RGD1565834; RP5-842K24.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55796 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NUK0 | |
Mbnl3 | |
Recombinant protein corresponding to Human MBNL3. Recombinant protein control fragment (Product #RP-92116). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.