Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBOAT1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
Antigen | MBOAT1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
MBOAT1 Polyclonal specifically detects MBOAT1 in Human samples. It is validated for Western Blot.Specifications
MBOAT1 | |
Polyclonal | |
Rabbit | |
Q6ZNC8 | |
154141 | |
Synthetic peptides corresponding to MBOAT1(membrane bound O-acyltransferase domain containing 1) The peptide sequence was selected from the N terminal of MBOAT1. Peptide sequence AAEPQPSSLSYRTTGSTYLHPLSELLGIPLDQVNFVVCQLVALFAAFWFR. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
dJ434O11.1, EC 2.3.1.-, EC 2.3.1.n6,1-acylglycerophosphoserine O-acyltransferase, LPEAT1, LPLAT 1, LPSAT, lysophosphatidylethanolamine acyltransferase 1, Lysophosphatidylserine acyltransferase, lysophospholipid acyltransferase 1, Lyso-PS acyltransferase, membrane bound O-acyltransferase domain containing 1, Membrane-bound O-acyltransferase domain-containing protein 1, MGC44669, OACT1, O-acyltransferase (membrane bound) domain containing 1, O-acyltransferase domain-containing protein 1 | |
MBOAT1 | |
IgG | |
56 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title