Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBOAT2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548906

Catalog No. PA548906

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Acyltransferase which mediates the conversion of lysophosphatidylethanolamine (1-acyl-sn-glycero-3- phosphoethanolamine or LPE) into phosphatidylethanolamine (1,2- diacyl-sn-glycero-3-phosphoethanolamine or PE) (LPEAT activity). Catalyzes also the acylation of lysophosphatidic acid (LPA) into phosphatidic acid (PA) (LPAAT activity). Has also a very weak lysophosphatidylcholine acyltransferase (LPCAT activity). Prefers oleoyl-CoA as the acyl donor. Lysophospholipid acyltransferases (LPLATs) catalyze the reacylation step of the phospholipid remodeling pathway also known as the Lands cycle.
Specifications

Specifications

MBOAT2
Polyclonal
PBS with 0.09% sodium azide
Q6ZWT7
Lysophospholipid acyltransferase 2, LPLAT 2, 231-, 1-acylglycerophosphate O-acyltransferase, 1-acylglycerophosphoethanolamine O-acyltransferase, 231n7, Lysophosphatidic acid acyltransferase, LPAAT, Lyso-PA acyltransferase, Lysophosphatidylethanolamine acyltransferase, LPEAT, Lyso-PE acyltransferase, Membrane-bound O-acyltransferase domain-containing protein 2, O-acyltransferase domain-containing protein 2, MBOAT2, OACT2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
129642
Western Blot
Unconjugated
MBOAT2
Liquid
MBOAT2
KLH conjugated synthetic peptide between 198-227 amino acids from the Central region of human MBOAT2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit