Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MBOAT4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565384
Description
Immunogen sequence: LDDSLLHAAGF GPELGQSPGE EGYVPDADIW TLERTHRISV FSRKWNQSTA RWLRRLVFQH S Highest antigen sequence identity to the following orthologs - mouse 81%, rat 79%.
Mediates the octanoylation of ghrelin at 'Ser-3'. Can use a variety of fatty acids as substrates including octanoic acid, decanoic acid and tetradecanoic acid.
Specifications
MBOAT4 | |
Polyclonal | |
Unconjugated | |
MBOAT4 | |
EMBL:ACB05875.1}; FKSG89; Ghrelin O-acyltransferase; ghrelin O-acyltransferase {ECO:0000303; Gm171; GOAT; Mboat4; mboat4 {ECO:0000312; membrane bound O-acyltransferase domain containing 4; membrane-bound O-acyltransferase domain-containing protein 4; membrane-bound O-acyltransferase domain-containing protein 4 {ECO:0000312; OACT4; O-acyltransferase (membrane bound) domain containing 4; O-acyltransferase domain-containing protein 4; PubMed:18443287}; RGD:1566335}; RGD1566335 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
619373 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96T53 | |
MBOAT4 | |
Recombinant Human MBOAT4. Recombinant protein control fragment (Product #RP-106070). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction