Learn More
MBP Antibody (MBP14), DyLight 488, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP261938G
Catalog No. NBP261938G
Add to Cart
Description
Description
MBP Monoclonal specifically detects MBP in Human, Bovine, Chicken, Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
MBP
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
MBP
Reacts with human MBP, recognizing epitopes in the 36-50 amino acid region (GILDSIGRFFGGDRG)
0.1 mL
Primary
Bovine, Chicken, Equine, Human
Purified
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
MBP14
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein
Rat
Protein G purified
Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers
4155
Store at 4°C in the dark.
IgG2b
Recommendations
Recommendations