Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBP Rabbit anti-Human, Clone: MBP/4277R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen MBP
Clone MBP/4277R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122026
View Documents
Novus Biologicals
NBP30727620UG
20 μg
Each for $159.82
Add to cart
 
NB122025
View Documents
Novus Biologicals
NBP30727610UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

MBP Monoclonal antibody specifically detects MBP in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

MBP
0.2 mg/ml
Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
Human
MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein
Recombinant fragment (around aa 150-250) of human MBP (exact sequence is proprietary)
Monoclonal
Protein A or G purified
MBP/4277R
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Unconjugated
Rabbit
Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers
10 mM PBS with 0.05% BSA
4155
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit