Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBP Rat anti-Human, Bovine, Chicken, Equine, Biotin, Clone: MBP14, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261938B

Catalog No. NBP261938B


Add to cart

Description

Description

MBP Monoclonal antibody specifically detects MBP in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

MBP
MBP14
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
MBP
Reacts with human MBP, recognizing epitopes in the 36-50 amino acid region (GILDSIGRFFGGDRG)
Protein G purified
Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers
Primary
4155
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Biotin
PBS with 0.05% Sodium Azide
MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein
Rat
IgG2b
0.1mL
Store at 4°C in the dark.
Monoclonal
Bovine, Chicken, Equine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit