Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBP Rat anti-Human, Bovine, Chicken, Equine, DyLight 405, Clone: MBP14, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261938V
Description
MBP Monoclonal antibody specifically detects MBP in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
MBP | |
MBP14 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
50mM Sodium Borate with 0.05% Sodium Azide | |
4155 | |
Rat | |
IgG2b | |
Primary | |
0.1mL | |
Store at 4°C in the dark. |
ELISA,Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Western Blot | |
DyLight 405 | |
Purified | |
MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
MBP | |
Reacts with human MBP, recognizing epitopes in the 36-50 amino acid region (GILDSIGRFFGGDRG) | |
Monoclonal | |
Protein G purified | |
Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers | |
Bovine,Chicken,Equine,Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title