Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MBP Rat anti-Human, Bovine, Chicken, Equine, DyLight 650, Clone: MBP14, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261938C
Description
MBP Monoclonal antibody specifically detects MBP in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
MBP | |
MBP14 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Purified | |
MBP | |
Reacts with human MBP, recognizing epitopes in the 36-50 amino acid region (GILDSIGRFFGGDRG) | |
Protein G purified | |
Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers | |
Primary | |
4155 |
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
Rat | |
IgG2b | |
0.1mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Bovine, Chicken, Equine, Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title