Learn More
MBP Rat anti-Human, Bovine, Chicken, Equine, DyLight 680, Clone: MBP14, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP261938FR
Catalog No. NBP261938FR
Add to cart
Description
Description
MBP Monoclonal antibody specifically detects MBP in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
MBP
MBP14
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
PBS with 0.05% Sodium Azide
4155
Rat
IgG2b
Primary
0.1mL
Store at 4°C in the dark.
ELISA,Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Western Blot
DyLight 680
Purified
MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein
MBP
Reacts with human MBP, recognizing epitopes in the 36-50 amino acid region (GILDSIGRFFGGDRG)
Monoclonal
Protein G purified
Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers
Bovine,Chicken,Equine,Human