Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBP-Tag Rabbit anti-Tag, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150891AP

Catalog No. 50-172-9969


Add to cart

Description

Description

This antibody recognizes MBP (Maltose binding protein) TAG in some expression systems.

The approach of epitope tagging offers an easy and universal strategy for the identification and purification of proteins derived by recombit DNA technology. The insertion of a Maltose Binding Protein (MBP) tag creates a stable fusion product that does not interfere with the bioactivity of the protein or with the biodistribution of the MBP-tagged product. Cleavage by factor Xa separates MBP from its partner protein.
Specifications

Specifications

MBP-Tag
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Maltose Binding Protein, MBP, MBP Tag
MBP-Tag Fusion Protein Ag0942
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Western Blot
Unconjugated
Liquid
Rabbit
IgG
150 μL
-20°C
Primary
Tag
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit