Learn More
MC1R Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 264711AP
Catalog No. 50-114-2744
Add to cart
Description
Description
MC1R, a Melanocortin Receptor, mediates the effects of melanocyte-stimulating hormone (MSH) and adrenocorticotropic hormone (ACTH). MSH and its receptor, MC1R, are key regulators of melanogenesis. They control the relative proportions of red pheomelanin and black photoprotective eumelanin in the skin by increasing the production of eumelanin. Non-functional, truncated forms of the receptor lead to lighter coat color in animals. In contrast, constitutively active receptors lead to darker color.
Specifications
Specifications
MC1R
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q01726
MC1 R, MC1R, Melanocortin receptor 1, MSH R, MSHR, SHEP2
Rabbit
IgG
150 μL
-20°C
Primary
4157
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
MC1R
Liquid
MC1R
MC1R Fusion Protein Ag24006
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human