Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MC3R Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
$496.07
Specifications
Antigen | MC3R |
---|---|
Concentration | 0.31 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3212
|
Proteintech Group Inc
210271AP150UL |
150 μL |
Each of 1 for $496.07
|
|
|||||
Description
This gene encodes a G-protein-coupled receptor for melanocyte-stimulating hormone and adrenocorticotropic hormone that is expressed in tissues other than the adrenal cortex and melanocytes. This gene maps to the same region as the locus for benign neonatal epilepsy. Mice deficient for this gene have increased fat mass despite decreased food intake, suggesting a role for this gene product in the regulation of energy homeostasis. Mutations in this gene are associated with a susceptibility to obesity in humans.Specifications
MC3R | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P41968 | |
4159 | |
MC3R Fusion Protein Ag15292 | |
Primary | |
-20°C | |
MC3R |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BMIQ9, MC3, MC3 R, MC3R, melanocortin 3 receptor, Melanocortin receptor 3, OB20, OQTL | |
MC3R | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title