Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCAM/CD146 Mouse anti-Human, FITC, Clone: 6C3E6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237365F

Catalog No. NBP237365F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

MCAM/CD146
6C3E6
Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
MCAM
Purified recombinant fragment of human MCAM (AA: 84-189) expressed in E. Coli.
Protein A or G purified
Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary
4162
ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
FITC
PBS with 0.05% Sodium Azide
CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18Gicerin
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit