Learn More
MCAM/CD146 Mouse anti-Rat, APC, Clone: 404722, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB3250A100UG
Catalog No. FB3250A100U
Add to cart
This item is not returnable. View return policy
Description
Description
MCAM/CD146 Monoclonal specifically detects MCAM/CD146 in Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
MCAM/CD146
Monoclonal
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, L-Gicerin, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18, MUC18Gicerin
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects rat MCAM/CD146 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) MCAM, rhBCAM, or rhALCAM is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG2a
Flow Cytometry
404722
Flow Cytometry 0.25-1 μg/106 cells
Q9EPF2
MCAM
Mouse myeloma cell line NS0-derived recombinant rat MCAM/CD146, Arg19-Lys560, Accession No. Q9EPF2
100 μg
4162.0
Rat
Supernatant