Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCAM/CD146 Rat anti-Mouse, Alexa Fluor 594, Clone: 733216, R&D Systems™

Rat MonoclonalAntibody

Manufacturer:  R&D Systems FAB7718T100UG

Catalog No. FAB7718T100


Add to cart
This item is not returnable. View return policy

Description

Description

MCAM/CD146 Monoclonal antibody specifically detects MCAM/CD146 in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

MCAM/CD146
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q8R2Y2
MCAM
Mouse myeloma cell line NS0-derived recombinant mouse MCAM/CD146 Met1-Val563 Accession # Q8R2Y2
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse MCAM/CD146 in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse (rm) MAdCAM-1 is observed, and no cross-reactivity with rmALCAM, rmNCAM, rmL1CAM, rmOCAM, rmTROP-2, recombinant human MCAM, or recombinant rat MCAM is observed.
Flow Cytometry
733216
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, L-Gicerin, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18, MUC18Gicerin
Rat
IgG2a
RUO
Primary
4162
Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)