Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCAM Monoclonal Antibody (OTI1F10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Supplier: OriGene TA803469
Description
CD146 (MCAM) is an integral membrane glycoprotein belonging to the immunoglobulin superfamily. CD146 is expressed at cell-cell junctions in all endothelial cells, and has also been observed in other normal and maligt cell types. In addition to its recognized role as a calcium-independent cell adhesion molecule, CD146 may also act as a signal transduction molecule in the recruitment of Fyn kinase, and in the subsequent tyrosine-phosphorylation of intracellular proteins involved in actin cytoskeleton assembly. CD146 plays a role in cell adhesion, and in cohesion of the endothelial monolayer at intercellular junctions in vascular tissue. CD146 expression may allow melanoma cells to interact with cellular elements of the vascular system, thereby enhancing hematogeneous tumor spread. Further, CD146 could be an adhesion molecule active in neural crest cells during embryonic development. CD146 acts also as surface receptor that triggers tyrosine phosphorylation of FYN and PTK2, and a transient increase in the intracellular calcium concentration.Specifications
MCAM | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P43121 | |
MCAM | |
Full length human recombit protein of human MCAM produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI1F10 | |
Unconjugated | |
MCAM | |
CD146, MUC18 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4162 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction