Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCAT Mouse anti-Human, DyLight™ 488, Clone: OTI2B7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272596G
Description
MCAT Monoclonal specifically detects MCAT in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
MCAT | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MCAT | |
Full length recombinant protein of human MCAT (NP_775738) produced in HEK293T cells. | |
0.1 mL | |
27349.0 | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2B7 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
EC 2.3.1.39, fabD, FASN2C, malonyl CoA:ACP acyltransferase (mitochondrial), malonyl-CoA:acyl carrier protein transacylase, mitochondrial, malonyl-CoA-acyl carrier protein transacylase, mitochondrial, MCT[Acyl-carrier-protein] malonyltransferase, Mitochondrial malonyltransferase, MTMGC47838, NET62 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title