Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCAT Polyclonal Antibody
MCAT Polyclonal Antibody
Supplier: Thermo Scientific PA564890
Description
MCAT Polyclonal Antibody for ICC/IF

Specifications
MCAT | |
Polyclonal | |
Unconjugated | |
MCAT | |
[Acyl-carrier-protein] malonyltransferase; [acyl-carrier-protein] S-malonyltransferase; AI225907; BC025519; fabD; FASN2C; malonyl CoA:ACP acyltransferase (mitochondrial); malonyl CoA-acyl carrier protein transacylase, mitochondrial; malonyl-CoA:acyl carrier protein transacylase, mitochondrial; malonyl-CoA-acyl carrier protein transacylase; malonyl-CoA-acyl carrier protein transacylase, mitochondrial; Mcat; MCT; mitochondrial malonyl CoA:ACP acyltransferase; Mitochondrial malonyltransferase; Mt; NET62; RGD1306698 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27349 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IVS2 | |
MCAT | |
Recombinant Human MCAT. Recombinant protein control fragment (Product #RP-105691). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction