Learn More
Apexbio Technology LLC MCC950 sodium(Synonyms: CP-456773 sodium, CRID3 sodium, MCC-950 sodium, MCC950 sodium salt), 50mg, CAS: 256373-96-3.
SDP
Supplier:  Apexbio Technology LLC B794650
Encompass_Preferred
MCC950 sodium (CAS 256373-96-3) is a selective small-molecule inhibitor targeting the NOD-like receptor family protein 3 (NLRP3) inflammasome MCC950 sodium inhibits NLRP3 activation in murine bone marrow-derived macrophages (BMDMs) with an IC50 of 7 5 nM and demonstrates comparable selectivity and potency in human monocyte-derived macrophages (HMDMs) It specifically blocks both canonical and noncanonical activation pathways without affecting other inflammasomes such as AIM2 NLRC4 and NLRP1 Due to its selectivity MCC950 sodium is valuable in investigating NLRP3-associated inflammatory and autoimmune conditions
Catalog No. 50-000-60878
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.