Leica Microsystems Paraplast™ X-tra

Tissue embedding media that clears completely without stainable residue

Manufacturer: Leica Microsystems 39501006

 View more versions of this product

Catalog No. 12-646-106

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Melting Point 56°C
    Special Use Minimizes tissue compression and wrinkling
    Product Type Paraplast™ Tissue Embedding Medium
    Quantity 1kg,1kg
    Type Paraplast™ Tissue Embedding Medium

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit