Learn More
Sklar InstrumentsSupplier Diversity Partner McCrea Urethral Sound, Sklar, 7 in., Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 852426
McCrea Urethral Sound, Sklar, 7 in., Stainless Steel. Sklar's McCrea X-Small Urethral Sound is commonly used with infants and female patients for dilation of the urethra to locate obstructions. This probe is 26 French (8.6mm) with a slightly curved tip.
Catalog No. 50-143-6434