Learn More
BioworldSupplier Diversity Partner MCDB 131 Medium w/o L-Glut, 500 mL DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 306110571
MCDB 131 Media were developed for the culture of specific cell types without a serum supplement. The media were supplemented with growth factors, hormones, trace elements or low levels of dialyzed fetal bovine serum protein (FBSP). Each MCDB medium was formulated for a specific cell type and was orginally developed for the clonal growth of human micro-vascular endothelial cells (HMVEC).
Catalog No. 50-254-137