Learn More
StatLabSupplier Diversity Partner MCDONALD'S GRAM STAIN KIT 1EA DFS Item
Manufacturer:  StatLabSupplier Diversity Partner KTMGS
This kit stains for gram positive and gram negative bacteria in less than five minutes. Stain kit includes Gentian Violet, Universal Iodine Solution, Gram's Decolorizer, Carbol Fuchsin, and Optimized Tartrazine Counterstain.
Catalog No. NC0180582