Learn More
MCF2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA548397
Catalog No. PA548397
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Guanine nucleotide exchange factor (GEF) that modulates the Rho family of GTPases. Promotes the conversion of some member of the Rho family GTPase from the GDP-bound to the GTP-bound form. Isoform 1 exhibits no activity toward RHOA, RAC1 or CDC42. Isoform 2 exhibits decreased GEF activity toward CDC42. Isoform 3 exhibits a weak but significant activity toward RAC1 and CDC42. Isoform 4 exhibits significant activity toward RHOA and CDC42. The truncated DBL oncogene is active toward RHOA, RAC1 and CDC42.
Specifications
Specifications
MCF2
Polyclonal
Unconjugated
MCF2
Liquid
MCF2
KLH conjugated synthetic peptide between 804-833 amino acids from the C-terminal region of human MCF2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.45 mg/mL
PBS with 0.09% sodium azide
P10911
Proto-oncogene DBL, Proto-oncogene MCF-2, MCF2-transforming protein, DBL-transforming protein, MCF2, DBL
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4168